Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026194.4 | NP_080470.2 | 9 | 937 | ||
GenBank mRNA | AK007993.2 | - | 9 | 882 | ||
AK013382.1 | - | 2 | 200 | |||
AK032663.1 | - | 9 | 873 | |||
AK129211.1 | - | 8 | 850 | |||
AK132697.1 | - | 9 | 901 | |||
BC058809.1 | - | 8 | 2261 | |||
BC080203.1 | - | 9 | 915 | |||
BC099529.1 | - | 9 | 924 | |||
BC132195.1 | - | 9 | 889 | 40130472 | ||
BC138153.1 | - | 9 | 889 | 8861172 |