Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Ahi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026203.3 | NP_080479.2 | 11 | 2090 | ||
GenBank mRNA | AK036336.1 | - | 11 | 1980 | ||
AY133241.1 | - | 11 | 1983 | |||
BC038502.1 | - | 4 | 441 | |||
BC055400.1 | - | 10 | 1812 | 6467369 | ||
BC065146.1 | - | 11 | 1986 |