Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026268.3 | NP_080544.1 | 1, 2 | 869 | ||
GenBank mRNA | AK005062.1 | - | 1, 2 | 870 | ||
AK009131.1 | - | 1, 2 | 797 | |||
AK088468.1 | - | 1, 2 | 870 | |||
AK088665.1 | - | 1, 2 | 871 | |||
AK151126.1 | - | 1, 2 | 866 | |||
AK152773.1 | - | 1, 2 | 866 | |||
AK159146.1 | - | 1, 2 | 770 | |||
AK159502.1 | - | 1, 2 | 870 | |||
AK159900.1 | - | 1, 2 | 770 | |||
BC003869.1 | - | 1, 2 | 451 | 3491528 |