Search Thermo Fisher Scientific
Gene Symbol: GCDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000159.3 | NP_000150.1 | 12 | 1831 | ||
NM_013976.3 | NP_039663.1 | 12 | 1599 | |||
NR_102316.1 | 12 | 1886 | ||||
NR_102317.1 | 11 | 2104 | ||||
GenBank mRNA | AK098370.1 | - | 12 | 1799 | ||
AK296446.1 | - | 12 | 1849 | |||
BC002579.2 | - | 12 | 1784 | 3142978 | ||
BM855466.1 | - | 1 | 442 | |||
U69141.1 | - | 11 | 1759 |