Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026298.5 | NP_080574.5 | 47 | 5225 | ||
GenBank mRNA | AK006007.1 | - | 4 | 502 | ||
AK016104.1 | - | 4 | 489 | |||
AK122461.1 | - | 28 | 5793 | |||
AY339616.1 | - | 47 | 5161 | |||
BC042583.1 | - | 47 | 5164 | |||
BC051928.1 | - | 8 | 723 | |||
BC060948.1 | - | 503 | ||||
BC066096.1 | - | 47 | 5218 | 6856438 |