Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2310057J18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026336.3 | NP_080612.1 | 1, 2 | 323 | ||
GenBank mRNA | AK008407.1 | - | 1, 2 | 321 | ||
AK009183.1 | - | 1, 2 | 322 | |||
AK009278.1 | - | 1, 2 | 321 | |||
AK009369.1 | - | 1, 2 | 320 | |||
AK009472.1 | - | 1, 2 | 321 | |||
AK009516.1 | - | 1, 2 | 321 | |||
AK009574.1 | - | 1, 2 | 320 | |||
AK009594.1 | - | 1, 2 | 321 | |||
AK009610.1 | - | 1, 2 | 321 | |||
AK009872.1 | - | 1, 2 | 322 | |||
AK009894.1 | - | 1, 2 | 352 | |||
AK009911.1 | - | 1, 2 | 322 | |||
AK009921.1 | - | 1, 2 | 321 | |||
AK009961.1 | - | 1, 2 | 324 | |||
AK010028.1 | - | 1, 2 | 321 | |||
AK010037.1 | - | 1, 2 | 324 | |||
AK010047.1 | - | 1, 2 | 319 | |||
AK010108.1 | - | 1, 2 | 321 | |||
AK010222.1 | - | 1, 2 | 323 | |||
AK010978.1 | - | 1, 2 | 324 | |||
AK075881.1 | - | 1, 2 | 320 | |||
AK075901.1 | - | 1, 2 | 32 | |||
BC021783.1 | - | 1, 2 | 308 | 4458723 | ||
BC055432.1 | - | 1, 2 | 310 |