Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VRK2 SIRNA Assays ›
Gene Symbol:
VRK2
Gene Name:
vaccinia related kinase 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 57907651 - 58163991 on Build GRCh38
UniGene ID:
Hs.715298
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
803
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VRK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130480.2
NP_001123952.1
5, 6
529
NM_001130481.2
NP_001123953.1
5, 6
371
NM_001130482.2
NP_001123954.1
5, 6
475
NM_001130483.2
NP_001123955.1
5, 6
529
NM_001288836.1
NP_001275765.1
6, 7
666
NM_001288837.1
NP_001275766.1
8, 9
1089
NM_001288838.1
NP_001275767.1
7, 8
971
NM_001288839.1
NP_001275768.1
4, 5
506
NM_006296.6
NP_006287.2
5, 6
576
XM_005264540.4
XP_005264597.1
5, 6
589
XM_006712090.3
XP_006712153.1
5, 6
384
XM_006712091.3
XP_006712154.1
5, 6
624
XM_006712092.3
XP_006712155.1
6, 7
761
XM_011533092.2
XP_011531394.1
5, 6
373
XM_017004858.1
XP_016860347.1
5, 6
529
XM_017004859.1
XP_016860348.1
6, 7
1251
XM_017004860.1
XP_016860349.1
5, 6
1181
XM_017004863.1
XP_016860352.1
5, 6
673
XM_017004864.1
XP_016860353.1
6, 7
761
XM_017004865.1
XP_016860354.1
6, 7
575
XM_017004866.1
XP_016860355.1
6, 7
624
GenBank mRNA
AB000450.1
-
5, 6
474
AJ512204.2
-
5, 6
474
AK058199.1
-
5, 6
431
AK223540.1
-
5, 6
431
AK292727.1
-
7, 8
957
AK295193.1
-
5, 6
421
AK296611.1
-
6, 7
651
AY228367.1
-
5, 6
371
AY228368.1
-
5, 6
506
AY228369.1
-
5, 6
404
AY228370.1
-
6, 7
596
AY228371.1
-
4, 5
436
AY228372.1
-
5, 6
459
BC021663.1
-
5, 6
402
BC027854.1
-
5, 6
527
5221315
BI906749.1
-
5, 6
536
DB227029.1
-
5, 6
529
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
regulation of cell shape
viral process
peptidyl-serine phosphorylation
cellular response to oxidative stress
regulation of MAPK cascade
protein autophosphorylation
regulation of interleukin-1-mediated signaling pathway
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0