Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem179b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026325.3 | NP_080601.2 | 4 | 502 | ||
GenBank mRNA | AK002731.1 | - | 2 | 157 | ||
AK005135.1 | - | 4 | 421 | |||
AK007906.1 | - | 4 | 478 | |||
AK010996.1 | - | 4 | 483 | |||
BC061068.1 | - | 4 | 402 | |||
BC087914.1 | - | 4 | 418 | |||
BC116961.1 | - | 4 | 502 | 40126280 | ||
BC116963.1 | - | 4 | 502 | 40126282 | ||
BC144813.1 | - | 4 | 432 |