Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294281.1 | NP_001281210.1 | 4, 5 | 429 | ||
NM_026350.3 | NP_080626.1 | 4, 5 | 429 | |||
GenBank mRNA | AK015911.1 | - | 4, 5 | 388 | ||
BC021321.1 | - | 3, 4 | 385 | 5041100 |