Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026428.2 | NP_080704.2 | 5 | 445 | ||
GenBank mRNA | AK002803.1 | - | 5 | 415 | ||
AK004023.1 | - | 5 | 444 | |||
AK007627.1 | - | 5 | 457 | |||
AK153521.1 | - | 5 | 426 | |||
AK161732.1 | - | 401 | ||||
AK178061.1 | - | 3 | 249 | |||
D89656.1 | - | 5 | 393 |