Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026438.4 | NP_080714.2 | 2 | 191 | ||
GenBank mRNA | AK008575.1 | - | 2 | 192 | ||
AK020291.1 | - | 2 | 195 | |||
AK033434.1 | - | 2 | 192 | |||
AK088397.1 | - | 2 | 187 | |||
AK145640.1 | - | 2 | 241 | |||
AK146300.1 | - | 2 | 154 | |||
AK151512.1 | - | 2 | 182 | |||
BC010468.1 | - | 2 | 125 | 3585780 | ||
CT010203.1 | - | 2 | 84 |