Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141965.1 | NP_001135437.1 | 5 | 472 | ||
NM_026425.3 | NP_080701.1 | 5 | 502 | |||
GenBank mRNA | AK005143.1 | - | 5 | 475 | ||
AK009229.1 | - | 5 | 474 | |||
AK153145.1 | - | 5 | 484 | |||
BC009157.1 | - | 5 | 439 | 2648626 | ||
BC096451.1 | - | 5 | 426 | 3673241 | ||
BC116374.2 | - | 5 | 431 | 40101000 | ||
CO802916.1 | - | 1 | 15 |