Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026454.3 | NP_080730.1 | 5 | 374 | ||
GenBank mRNA | AK007549.1 | - | 5 | 345 | ||
AK007937.1 | - | 5 | 374 | |||
AK007960.1 | - | 5 | 364 | |||
AK010957.1 | - | 6 | 451 | |||
AK030594.1 | - | 354 | ||||
AK153481.1 | - | 5 | 375 | |||
AK208168.1 | - | 4 | 158 | |||
BC016117.1 | - | 6 | 656 | 4486610 |