Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294281.1 | NP_001281210.1 | 9 | 1681 | ||
NM_026350.3 | NP_080626.1 | 10 | 1820 | |||
GenBank mRNA | AK015911.1 | - | 10 | 1779 |