Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tprgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026388.2 | NP_080664.1 | 4 | 1353 | ||
GenBank mRNA | AK004324.1 | - | 3 | 966 | ||
AK004777.1 | - | 4 | 1334 | |||
AK155413.1 | - | 4 | 1348 | |||
AK185356.1 | - | 1 | 293 | |||
AK190435.1 | - | 1 | 212 | |||
AK193406.1 | - | 1 | 212 | |||
AK206227.1 | - | 1 | 211 | |||
AK210830.1 | - | 1 | 212 | |||
AK211936.1 | - | 1 | 130 | |||
AK212048.1 | - | 1 | 211 | |||
AK212439.1 | - | 1 | 213 | |||
AK215133.1 | - | 1 | 211 | |||
AK215732.1 | - | 1 | 212 | |||
BC019544.1 | - | 4 | 1317 | 4211201 |