Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026391.2 | NP_080667.1 | 9 | 1717 | ||
GenBank mRNA | AF366393.1 | - | 9 | 1625 | ||
AK005033.1 | - | 8 | 1606 | |||
AK129386.2 | - | 9 | 1744 | |||
BC066022.1 | - | 9 | 1717 | 6828707 |