Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prorsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163454.2 | NP_001156926.1 | 3 | 517 | ||
NM_026465.3 | NP_080741.1 | 2 | 426 | |||
GenBank mRNA | AK008574.1 | - | 2 | 427 | ||
AK077280.1 | - | 3 | 515 | |||
AK085965.1 | - | 3 | 494 | |||
BC033930.1 | - | 3 | 490 | 1397170 | ||
BC115900.1 | - | 2 | 427 | 40091622 |