Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700062C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026529.4 | NP_080805.1 | 2, 3 | 143 | ||
GenBank mRNA | AK011228.1 | - | 1, 2 | 230 | ||
AK012467.1 | - | 1, 2 | 245 | |||
AK013030.1 | - | 2, 3 | 53 | |||
AK018391.1 | - | 2, 3 | 147 | |||
AK164298.1 | - | 1 | 1846 | |||
BC029246.1 | - | 2, 3 | 148 | |||
BC037075.1 | - | 2, 3 | 191 | 5038294 | ||
BC084681.1 | - | 2, 3 | 153 | 30144888 |