Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026530.5 | NP_080806.4 | 3 | 372 | ||
GenBank mRNA | AK012128.1 | - | 2 | 327 | ||
AK146973.1 | - | 3 | 357 | |||
AK160356.1 | - | 3 | 341 | |||
AK172381.1 | - | 2 | 115 | |||
AK199908.1 | - | 2 | 90 | |||
BC017625.1 | - | 2 | 321 | |||
CJ180905.1 | - | 3 | 374 |