Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168471.1 | NP_001161943.1 | 2 | 181 | ||
NM_001168472.1 | NP_001161944.1 | 2 | 354 | |||
NM_026556.4 | NP_080832.1 | 2 | 387 | |||
GenBank mRNA | AK006876.1 | - | 2 | 163 | ||
AK011284.1 | - | 2 | 355 | |||
AK028319.1 | - | 2 | 152 | |||
AK049645.1 | - | 2 | 361 | |||
AK078434.1 | - | 2 | 360 | |||
AK160114.1 | - | 2 | 357 | |||
AK169212.1 | - | 2 | 360 | |||
AY029255.1 | - | 1 | 89 | |||
BC011289.1 | - | 2 | 178 | 3981362 | ||
BC040822.1 | - | 2 | 349 | 4459858 | ||
BC049621.1 | - | 160 | ||||
BY249698.1 | - | 2 | 354 |