Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B230118H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026592.3 | NP_080868.1 | 2, 3 | 268 | ||
GenBank mRNA | AK010820.1 | - | 2, 3 | 231 | ||
AK020980.1 | - | 2, 3 | 269 | |||
AK051679.1 | - | 2, 3 | 269 | |||
AK088637.1 | - | 2, 3 | 312 | |||
BC025075.1 | - | 2, 3 | 272 | 4037434 | ||
DQ112091.1 | - | 481 | ||||
DQ112092.1 | - | 273 |