Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam135a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026604.4 | NP_080880.4 | 14 | 1834 | ||
GenBank mRNA | AK034175.1 | - | 1 | 200 | ||
AK053786.1 | - | 14 | 1753 | |||
AK149303.1 | - | 14 | 1826 | |||
BC070446.1 | - | 13 | 1516 | 30362784 |