Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026497.2 | NP_080773.1 | 2, 3 | 324 | ||
GenBank mRNA | AK002641.1 | - | 2, 3 | 325 | ||
AK083846.1 | - | 2, 3 | 282 | |||
AK147867.1 | - | 2, 3 | 278 | |||
BC057657.1 | - | 2, 3 | 278 | 2646355 |