Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026578.3 | NP_080854.1 | 5, 6 | 909 | ||
GenBank mRNA | AK021256.1 | - | 5, 6 | 670 | ||
AK075923.1 | - | 4, 5 | 644 | |||
AK167001.1 | - | 5, 6 | 649 | |||
BC021873.1 | - | 6, 7 | 773 | 3709271 | ||
BC048685.1 | - | 5, 6 | 828 | 6510731 |