Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700062C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026529.4 | NP_080805.1 | 5 | 583 | ||
GenBank mRNA | AK011228.1 | - | 4 | 670 | ||
AK012467.1 | - | 4 | 685 | |||
AK013030.1 | - | 5 | 491 | |||
AK018391.1 | - | 5 | 587 | |||
AK164298.1 | - | 3 | 2286 | |||
BC029246.1 | - | 5 | 1240 | |||
BC037075.1 | - | 5 | 631 | 5038294 | ||
BC084681.1 | - | 5 | 593 | 30144888 |