Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ebpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026598.3 | NP_080874.2 | 4 | 1044 | ||
GenBank mRNA | AK019963.1 | - | 4 | 1039 | ||
AK044618.1 | - | 2 | 1532 |