Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Coa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026618.2 | NP_080894.1 | 2 | 567 | ||
GenBank mRNA | AK003955.1 | - | 2 | 566 | ||
AK019025.1 | - | 2 | 569 | |||
AK178548.1 | - | 1 | 57 | |||
BC026206.1 | - | 2 | 561 | 1265058 |