Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700067P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026625.2 | NP_080901.2 | 3 | 351 | ||
GenBank mRNA | AK015495.1 | - | 3 | 313 | ||
AK018887.1 | - | 3 | 340 | |||
BC048440.1 | - | 3 | 341 | 6741836 |