Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026886.3 | NP_081162.1 | 2 | 529 | ||
GenBank mRNA | AK005085.1 | - | 2 | 529 | ||
AK053836.1 | - | 2 | 261 | |||
AK165719.1 | - | 2 | 247 | |||
BC056392.1 | - | 2 | 519 | 6855144 | ||
JN860221.1 | - | 2 | 176 |