Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026981.2 | NP_081257.1 | 2 | 280 | ||
GenBank mRNA | AK007678.1 | - | 1 | 247 | ||
AK149746.1 | - | 2 | 281 | |||
BC020149.1 | - | 2 | 227 | 3993232 | ||
BY240339.1 | - | 2 | 283 |