Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083895.3 | NP_001077364.2 | 3 | 595 | ||
NM_026838.4 | NP_081114.2 | 4 | 647 | |||
GenBank mRNA | AF393640.1 | - | 2 | 165 | ||
AK136935.1 | - | 3 | 450 | |||
BC028307.1 | - | 4 | 376 | 3488526 | ||
CN539245.1 | - | 4 | 410 |