Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026907.3 | NP_081183.3 | 4 | 503 | ||
GenBank mRNA | AF210700.1 | - | 3 | 406 | ||
AK007321.1 | - | 3 | 365 | |||
AK029082.1 | - | 4 | 504 | |||
AK143807.1 | - | 4 | 487 | |||
BC010805.1 | - | 4 | 580 | 4239879 | ||
BC020159.1 | - | 3 | 454 | 4016267 |