Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026917.5 | NP_081193.2 | 7 | 1004 | ||
GenBank mRNA | AB088044.1 | - | 6 | 770 | ||
AK007355.1 | - | 8 | 1476 | |||
AK018755.1 | - | 7 | 971 | |||
AK080541.1 | - | 7 | 970 | |||
AY166672.1 | - | 6 | 770 | |||
BC005531.1 | - | 8 | 1031 | |||
BC018334.1 | - | 6 | 840 | |||
BC025126.1 | - | 7 | 989 | 3669723 | ||
BC052464.1 | - | 7 | 909 | 6834763 |