Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271977.1 | NP_001258906.1 | 12 | 1673 | ||
NM_001271978.1 | NP_001258907.1 | 11 | 1645 | |||
NM_001271979.1 | NP_001258908.1 | 11 | 1442 | |||
NM_001271980.1 | NP_001258909.1 | 11 | 1380 | |||
NM_001282305.1 | NP_001269234.1 | 11 | 1640 | |||
NM_001282306.1 | NP_001269235.1 | 12 | 1623 | |||
NM_001282307.1 | NP_001269236.1 | 11 | 1543 | |||
NM_001282308.1 | NP_001269237.1 | 11 | 1481 | |||
NM_006374.4 | NP_006365.2 | 12 | 1611 | |||
XM_011510494.2 | XP_011508796.1 | 12 | 1476 | |||
XM_011510495.2 | XP_011508797.1 | 11 | 1748 | |||
XM_017003168.1 | XP_016858657.1 | 11 | 1550 | |||
XM_017003169.1 | XP_016858658.1 | 11 | 1431 | |||
XM_017003170.1 | XP_016858659.1 | 11 | 1379 | |||
XM_017003171.1 | XP_016858660.1 | 10 | 1351 | |||
GenBank mRNA | AK092161.1 | - | 11 | 1303 | ||
AK291808.1 | - | 11 | 1269 | |||
AK291947.1 | - | 12 | 1500 | |||
AK297374.1 | - | 11 | 1432 | |||
AK301214.1 | - | 12 | 1511 | |||
AK303719.1 | - | 11 | 1613 | |||
AK315966.1 | - | 11 | 1381 | |||
BC007852.2 | - | 12 | 1535 | 4298239 | ||
BC015793.1 | - | 11 | 1591 | 4867978 | ||
BC091505.1 | - | 12 | 1565 | 30383354 | ||
CR936874.1 | - | 2 | 4336 | |||
D63780.1 | - | 11 | 1434 | |||
X99325.1 | - | 11 | 1446 |