Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164607.1 | NP_001158079.1 | 9 | 1718 | ||
NM_001164608.1 | NP_001158080.1 | 8 | 1625 | |||
NM_026859.3 | NP_081135.3 | 8 | 1672 | |||
GenBank mRNA | AK004406.1 | - | 8 | 1345 | ||
AK042913.1 | - | 8 | 1566 | |||
AK154047.1 | - | 8 | 1615 | |||
BB250147.2 | - | 2 | 630 | |||
BC016260.1 | - | 8 | 1660 | 4914167 | ||
BY595681.1 | - | 1 | 424 |