Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cd209b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037800.3 | NP_001032889.3 | 5, 6 | 435 | ||
NM_001287211.1 | NP_001274140.1 | 5, 6 | 435 | |||
NM_026972.5 | NP_081248.4 | 6, 7 | 525 | |||
GenBank mRNA | AF373409.1 | - | 6, 7 | 503 | ||
AF374471.1 | - | 5, 6 | 392 | |||
AF422108.1 | - | 6, 7 | 498 | |||
AF424790.1 | - | 6, 7 | 497 | |||
AF424791.1 | - | 6, 7 | 497 | |||
AF424792.1 | - | 6, 7 | 497 | |||
AF424793.1 | - | 6, 7 | 497 | |||
AF424794.1 | - | 6, 7 | 517 | |||
AF424795.1 | - | 6, 7 | 514 | |||
AF424796.1 | - | 6, 7 | 508 | |||
AF424797.1 | - | 5, 6 | 407 | |||
AF424798.1 | - | 5, 6 | 427 | |||
AF424799.1 | - | 5, 6 | 407 | |||
AF424803.1 | - | 6, 7 | 533 | |||
AK007656.1 | - | 5, 6 | 441 | |||
AK138999.1 | - | 6, 7 | 499 |