Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAMKK2 SIRNA Assays ›
Gene Symbol:
CAMKK2
Gene Name:
calcium/calmodulin dependent protein kinase kinase 2
Gene Aliases:
CAMKK, CAMKKB
Chromosome Location:
Chr.12: 121237692 - 121298308 on Build GRCh38
UniGene ID:
Hs.297343
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
815
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAMKK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270485.1
NP_001257414.1
2, 3
654
NM_001270486.1
NP_001257415.1
1, 2
538
NM_006549.3
NP_006540.3
2, 3
1308
NM_153499.2
NP_705719.2
2, 3
1308
NM_153500.1
NP_705720.1
2, 3
1308
NM_172214.2
NP_757363.1
2, 3
1308
NM_172215.2
NP_757364.1
2, 3
1308
NM_172216.1
NP_757365.1
2, 3
1308
NM_172226.2
NP_757380.1
2, 3
654
XM_005253822.3
XP_005253879.1
2, 3
610
XM_005253823.1
XP_005253880.1
2, 3
610
XM_005253824.3
XP_005253881.1
2, 3
610
XM_011537763.1
XP_011536065.1
2, 3
610
XM_011537764.2
XP_011536066.1
2, 3
610
XM_017018694.1
XP_016874183.1
2, 3
610
XM_017018695.1
XP_016874184.1
2, 3
819
XM_017018696.1
XP_016874185.1
2, 3
661
XM_017018697.1
XP_016874186.1
2, 3
683
XM_017018698.1
XP_016874187.1
2, 3
661
XM_017018699.1
XP_016874188.1
2, 3
610
XM_017018700.1
XP_016874189.1
2, 3
610
XM_017018701.1
XP_016874190.1
2, 3
661
XM_017018702.1
XP_016874191.1
2, 3
819
XM_017018703.1
XP_016874192.1
2, 3
610
XM_017018704.1
XP_016874193.1
2, 3
661
XM_017018705.1
XP_016874194.1
2, 3
610
XM_017018706.1
XP_016874195.1
2, 3
610
XM_017018707.1
XP_016874196.1
1, 2
533
XM_017018708.1
XP_016874197.1
2, 3
610
XM_017018709.1
XP_016874198.1
2, 3
613
GenBank mRNA
AB018330.2
-
1, 2
484
AB081336.1
-
1, 2
511
AB081337.1
-
1, 2
511
AF101264.1
-
1, 2
131
AF140507.1
-
2, 3
592
AF287630.1
-
2, 3
1302
AF287631.1
-
2, 3
1302
AF321385.1
-
2, 3
1058
AF321386.1
-
2, 3
1058
AF321387.1
-
2, 3
1058
AF321388.1
-
2, 3
1058
AK024748.1
-
1, 2
294
AK292072.1
-
2, 3
802
AK300865.1
-
3, 4
554
BC026060.2
-
2, 3
587
3842706
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031338, AB018081.
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein phosphorylation
peptidyl-tyrosine phosphorylation
calcium-mediated signaling
regulation of protein kinase activity
positive regulation of transcription, DNA-templated
protein autophosphorylation
calmodulin-dependent protein kinase activity
protein tyrosine kinase activity
calcium ion binding
calmodulin binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline