Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lce1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027137.2 | NP_081413.1 | 2 | 607 | ||
GenBank mRNA | AK009666.1 | - | 2 | 608 | ||
BC060282.1 | - | 2 | 579 |