Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027175.3 | NP_081451.3 | 2 | 342 | ||
GenBank mRNA | AK010328.1 | - | 1 | 172 | ||
AK050343.1 | - | 2 | 343 | |||
AK215302.1 | - | 1 | 113 | |||
BC018422.1 | - | 2 | 172 | 3593650 |