Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172055.1 | NP_001165526.1 | 6 | 613 | ||
NM_027208.2 | NP_081484.1 | 6 | 540 | |||
GenBank mRNA | AK007603.1 | - | 4 | 264 | ||
BC021461.1 | - | 6 | 617 | |||
BC036998.1 | - | 6 | 480 | 5041996 |