Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081009.1 | NP_001074478.1 | 27 | 2674 | ||
GenBank mRNA | AK013206.1 | - | 11 | 826 | ||
AK149889.1 | - | 26 | 2647 | |||
AK149918.1 | - | 26 | 2641 | |||
AK150208.1 | - | 26 | 2641 | |||
AK150732.1 | - | 25 | 2451 | |||
AK151510.1 | - | 25 | 2452 | |||
AK152919.1 | - | 25 | 2451 | |||
AK153169.1 | - | 25 | 2451 | |||
AK153270.1 | - | 25 | 2451 | |||
BC021315.1 | - | 17 | 1808 | 5027417 | ||
BC021881.1 | - | 26 | 2644 | 3710450 | ||
BC022679.1 | - | 12 | 1201 | |||
EF591877.1 | - | 2409 |