Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2010107E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027360.2 | NP_081636.1 | 2 | 128 | ||
GenBank mRNA | AK008333.1 | - | 2 | 129 | ||
AK020400.1 | - | 1 | 544 | |||
BC055712.1 | - | 2 | 107 | 6408438 | ||
BC059719.1 | - | 2 | 116 | 6774233 |