Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027422.2 | NP_081698.2 | 14 | 2138 | ||
GenBank mRNA | AK012576.1 | - | 2 | 986 | ||
AK154672.1 | - | 14 | 2155 | |||
AK173128.1 | - | 13 | 2293 | |||
BC005748.1 | - | 1 | 546 | |||
BC067013.1 | - | 14 | 2138 | 6810181 | ||
BC080757.1 | - | 7 | 1389 | |||
BC083321.1 | - | 5 | 1259 |