Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323011.1 | NP_001309940.1 | 2, 3 | 125 | ||
NM_001323012.1 | NP_001309941.1 | 2, 3 | 132 | |||
NM_006556.3 | NP_006547.1 | 2, 3 | 475 | |||
GenBank mRNA | AK130234.1 | - | 3, 4 | 317 | ||
AK312803.1 | - | 2, 3 | 434 | |||
BC006089.1 | - | 2, 3 | 458 | 3604167 | ||
BC007694.2 | - | 2, 3 | 388 | 3954909 | ||
BQ061044.1 | - | 2, 3 | 475 | |||
CR541998.1 | - | 2, 3 | 152 | |||
L77213.1 | - | 2, 3 | 187 |