Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027439.4 | NP_081715.1 | 2 | 224 | ||
GenBank mRNA | AB192471.2 | - | 2 | 100 | ||
AK017482.1 | - | 2 | 156 | |||
AK029405.1 | - | 2 | 127 | |||
AK076495.1 | - | 2 | 169 | |||
AK151735.1 | - | 2 | 155 | |||
AK168139.1 | - | 2 | 183 | |||
AK210696.1 | - | 1 | 43 | |||
BC014706.1 | - | 2 | 95 | 4457079 | ||
BY784908.1 | - | 2 | 225 |