Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027494.3 | NP_081770.3 | 4 | 470 | ||
GenBank mRNA | AK017857.1 | - | 4 | 366 | ||
AK032349.1 | - | 4 | 448 | |||
AK033469.1 | - | 4 | 420 | |||
AK156770.1 | - | 4 | 442 | |||
AK166990.1 | - | 4 | 377 | |||
AK167126.1 | - | 4 | 375 | |||
AK167736.1 | - | 4 | 377 | |||
BC013555.1 | - | 3 | 122 | 4238634 |