Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027570.3 | NP_081846.3 | 2 | 119 | ||
GenBank mRNA | AK014649.1 | - | 2 | 134 | ||
AK029820.1 | - | 133 | ||||
AY092768.1 | - | 2 | 124 | |||
BC039155.1 | - | 2 | 115 | 4242682 | ||
BC055443.1 | - | 2 | 119 | 4235345 |