Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243132.1 | NP_001230061.1 | 2 | 169 | ||
NM_027533.3 | NP_081809.2 | 2 | 364 | |||
GenBank mRNA | AK018178.1 | - | 2 | 167 | ||
AK020982.1 | - | 2 | 169 | |||
AK031683.1 | - | 1 | 73 | |||
AK032520.1 | - | 2 | 190 | |||
AK049888.1 | - | 2 | 166 | |||
AK079560.1 | - | 2 | 168 | |||
BB642061.1 | - | 2 | 365 | |||
BC007185.1 | - | 2 | 605 | 3591472 |