Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164336.1 | NP_001157808.1 | 3 | 640 | ||
NM_001164337.1 | NP_001157809.1 | 3 | 640 | |||
NM_027314.3 | NP_081590.3 | 3 | 640 | |||
GenBank mRNA | AK007511.1 | - | 3 | 476 | ||
AK009232.1 | - | 3 | 626 | |||
AK009364.1 | - | 3 | 638 | |||
AK021403.1 | - | 3 | 611 | |||
AK178944.1 | - | 2 | 154 | |||
AK206577.1 | - | 1 | 35 | |||
AK219813.1 | - | 2 | 102 | |||
BC064752.1 | - | 3 | 364 | 30049608 | ||
BC107214.1 | - | 3 | 351 | |||
BC107215.1 | - | 3 | 352 | 40051628 | ||
BC132455.1 | - | 3 | 404 | 40130732 | ||
BC132457.1 | - | 3 | 403 | 40130734 |