Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027462.4 | NP_081738.2 | 6 | 1016 | ||
GenBank mRNA | AK017602.1 | - | 6 | 1027 | ||
AK036902.1 | - | 7 | 1087 | |||
AK042715.1 | - | 6 | 1017 | |||
AK042964.1 | - | 6 | 1017 | |||
AK185879.1 | - | 1 | 159 | |||
BC089583.1 | - | 6 | 1038 | 6762967 |